Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Span® 80

Glass bottle for solids, 1 kg
Catalog # ROTH9426.2
Marketsource
Supplier:   Roth Carl
Span® 80
Catalog # ROTH9426.2
Supplier:  Roth Carl
Marketsource
CAS Number:  1338-43-8
Span® 80

Glass bottle for solids, 1 kg

CAS Number:  1338-43-8
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Arlacel 80, Sorbitan monooleate
  • CAS Number:1338-43-8
  • Formula:C₂₄H₄₄O₆
  • MDL Number:MFCD00080948
  • Molecular Weight:428.61 g/mol
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-665-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.