Order Entry
Denmark
Orders LinkContactUsLinkComponent
Acetone

Acetone

Glass bottle, 1 L
Katalog # ROTH9372.1
Flammable
Irritant
Marketsource
Leverandør:   Roth Carl
Acetone
Katalog # ROTH9372.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  67-64-1
Acetone
Acetone

Glass bottle, 1 L

CAS nummer:  67-64-1
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.