Order Entry
Denmark
Orders LinkContactUsLinkComponent
Imidazole

Imidazole

Glass bottle for solids, 250 g
Catalog # ROTHX998.4
Corrosive
Health
Irritant
Marketsource
Supplier:   Roth Carl
Imidazole
Catalog # ROTHX998.4
Supplier:  Roth Carl
Marketsource
CAS Number:  288-32-4
Imidazole
Imidazole

Glass bottle for solids, 250 g

CAS Number:  288-32-4
Price
Inquire For Price
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Merck Index:13,04935
  • Boiling Pt:255 °C (1013 hPa)
  • Melting Pt:89…90 °C
  • Density:1.18 g/cm³ (20 °C)
  • Flash Pt:135 °C
  • Storage Temperature:Ambient
  • EINECS:206-019-2
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.