Propylene oxide
Glass bottle, 2,5 L
ROTH8617.2
Marketsource
: (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
- :75-56-9
- :C₃H₆O
- :MFCD00005126
- :58.08 g/mol
- :12,08041
- :34.3 °C (1013 hPa)
- :–112 °C
- :0.83 g/cm³ (20 °C)
- :–37 °C
- :Refrigerator
- :200-879-2
- :1280
- :3,I
- Pack type:Glass bottle
- Pk:2,5 L