Sølvnitrat
50 g
Catalog # ROTH7908.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- CAS Number:7761-88-8
- Formula:AgNO₃
- MDL Number:MFCD00003414
- Molecular Weight:169.87 g/mol
- Merck Index:12,08661
- Boiling Pt:444 °C (1013 hPa)
- Melting Pt:210 °C
- Density:4,35 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:231-853-9
- UN:1493
- ADR:5.1,II
- REACH:01-2119513705-43
Specifications
Specifications
- Pakn:50 g



