Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Phenol

Glass bottle, 100 mL
Catalog # ROTH0038.1
Toxic
Corrosive
Health
Marketsource
Supplier:   Roth Carl
Phenol
Catalog # ROTH0038.1
Supplier:  Roth Carl
Marketsource
CAS Number:  108-95-2
Phenol

Glass bottle, 100 mL

CAS Number:  108-95-2
Price
Inquire For Price
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Merck Index:12,07390
  • Boiling Pt:181,7 °C (1013 hPa)
  • Melting Pt:40,8 °C
  • Density:1,0722 g/cm³ (20 °C)
  • Flash Pt:81 °C
  • Storage Temperature:Stue temperatur
  • EINECS:203-632-7
  • UN:1671
  • ADR:6.1,II
  • REACH:01-2119471329-32
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    100 mL

Specifications