Order Entry
Denmark
Orders LinkContactUsLinkComponent
Potassium acetate

Potassium acetate

Glass bottle for solids, 500 g
Catalog # ROTHT874.1
Marketsource
Supplier:   Roth Carl
Potassium acetate
Catalog # ROTHT874.1
Supplier:  Roth Carl
Marketsource
CAS Number:  127-08-2
Potassium acetate
Potassium acetate

Glass bottle for solids, 500 g

CAS Number:  127-08-2
Price
317,37 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Acetic acid potassium salt
  • CAS Number:127-08-2
  • Formula:CH₃COOK
  • MDL Number:MFCD00012458
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,07687
  • Melting Pt:304 °C
  • Density:1.57 g/cm³ (25 °C)
  • Flash Pt:>250 °C
  • Storage Temperature:Ambient
  • EINECS:204-822-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.