Order Entry
Denmark
Orders LinkContactUsLinkComponent
Agarose

Agarose

Glass bottle for solids, 10 g
Catalog # ROTHT846.1
Marketsource
Supplier:   Roth Carl
Agarose
Catalog # ROTHT846.1
Supplier:  Roth Carl
Marketsource
CAS Number:  9012-36-6
Agarose
Agarose

Glass bottle for solids, 10 g

CAS Number:  9012-36-6
Price
230,75 kr
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Ambient
  • EINECS:232-731-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.