Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzene-[D₆]

Benzene-[D₆]

Glass bottle, 50 mL
Catalog # ROTHAE53.2
Flammable
Health
Marketsource
Supplier:   Roth Carl
Benzene-[D₆]
Catalog # ROTHAE53.2
Supplier:  Roth Carl
Marketsource
CAS Number:  1076-43-3
Benzene-[D₆]

Glass bottle, 50 mL

CAS Number:  1076-43-3
Price
2.680,88 kr
Log in for availability
1 * 50 mL
Product Details & Documents
Synonyms:  Benzene-[²H₆]
  • CAS Number:1076-43-3
  • Formula:C₆D₆
  • MDL Number:MFCD00003010
  • Molecular Weight:84.07 g/mol
  • Boiling Pt:79 °C (1013 hPa)
  • Melting Pt:6.7 °C
  • Density:0.95 g/cm³ (20 °C)
  • Flash Pt:–11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:214-061-8
  • UN:1114
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL

Specifications