Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzene-[D₆]

Benzene-[D₆]

Glass bottle, 10 mL
Catalog # ROTHAE53.1
Flammable
Health
Marketsource
Supplier:   Roth Carl
Benzene-[D₆]
Catalog # ROTHAE53.1
Supplier:  Roth Carl
Marketsource
CAS Number:  1076-43-3
Benzene-[D₆]

Glass bottle, 10 mL

CAS Number:  1076-43-3
Price
671,34 kr
Log in for availability
1 * 10 mL
Product Details & Documents
Synonyms:  Benzene-[²H₆]
  • CAS Number:1076-43-3
  • Formula:C₆D₆
  • MDL Number:MFCD00003010
  • Molecular Weight:84.07 g/mol
  • Boiling Pt:79 °C (1013 hPa)
  • Melting Pt:6.7 °C
  • Density:0.95 g/cm³ (20 °C)
  • Flash Pt:–11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:214-061-8
  • UN:1114
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.