Order Entry
Denmark
Orders LinkContactUsLinkComponent
Guanine
Glass bottle for solids, 10 g
  ROTH7505.1
Marketsource
 :   Roth Carl
Guanine
  ROTH7505.1
 :  Roth Carl
Marketsource
 :  73-40-5
Guanine
Glass bottle for solids, 10 g
 :  73-40-5
1 * 10 g
 :  2-Amino-1H-purin-6(7H)-one
  •  :73-40-5
  •  :C₅H₅N₅O
  •  :MFCD00071533
  •  :151.13 g/mol
  •  :13,04580
  •  :> 220 °C
  •  :1.67 g/cm³ (20 °C)
  •  :Ambient
  •  :200-799-8
  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g