Order Entry
Denmark
Orders LinkContactUsLinkComponent
Acetonitrile

Acetonitrile

Glass bottle, 1 L
Catalog # ROTH7330.1
Flammable
Irritant
Marketsource
   Safer Human and Environmental Health
Supplier:   Roth Carl
Acetonitrile
Catalog # ROTH7330.1
Supplier:  Roth Carl
Marketsource
   Safer Human and Environmental Health
CAS Number:  75-05-8
Acetonitrile
Acetonitrile

Glass bottle, 1 L

CAS Number:  75-05-8
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
  • CAS Number:75-05-8
  • Formula:H₃CCN
  • MDL Number:MFCD00001878
  • Molecular Weight:41.05 g/mol
  • Merck Index:13,00071
  • Boiling Pt:81.6 °C (1013 hPa)
  • Melting Pt:–45.7 °C
  • Density:0.782 g/cm³ (20 °C)
  • Flash Pt:2 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-835-2
  • UN:1648
  • ADR:3,II
  • REACH:01-2119471307-38
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Cat. No.:
    ROTH7330.1
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.