Benzene
Glass bottle, 100 mL
Catalog # ROTH7173.4
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Cyclohexatriene
- CAS Number:71-43-2
- Formula:C₆H₆
- MDL Number:MFCD00003009
- Molecular Weight:78.11 g/mol
- Merck Index:13,01066
- Boiling Pt:80.1 °C (1013 hPa)
- Melting Pt:5.5 °C
- Density:0.878 g/cm³ (20 °C)
- Flash Pt:–11 °C
- Storage Temperature:Ambient
- EINECS:200-753-7
- UN:1114
- ADR:3,II
- REACH:01-2119447106-44
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL



