Tris acetate
Glass bottle for solids, 25 g
Catalog # ROTH7125.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
- CAS Number:6850-28-8
- Formula:NH₂C(CH₂OH)₃·CH₃COOH
- MDL Number:MFCD00038951
- Molecular Weight:181.1870 g/mol
- Storage Temperature:Ambient
- EINECS:229-939-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



