Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Formamide

Glass bottle, 2,5 L
Catalog # ROTH6749.2
Health
Marketsource
Supplier:   Roth Carl
Formamide
Catalog # ROTH6749.2
Supplier:  Roth Carl
Marketsource
CAS Number:  75-12-7
Formamide
Formamide

Glass bottle, 2,5 L

CAS Number:  75-12-7
Price
Inquire For Price
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:13,04261
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:2 °C
  • Density:1.1334 g/cm³ (20 °C)
  • Flash Pt:79 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-842-0
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications