Order Entry
Denmark
Orders LinkContactUsLinkComponent
Beeswax

Beeswax

Glass bottle for solids, 1 kg
Catalog # ROTH5825.2
Marketsource
Supplier:   Roth Carl
Beeswax
Catalog # ROTH5825.2
Supplier:  Roth Carl
Marketsource
CAS Number:  8012-89-3
Beeswax

Glass bottle for solids, 1 kg

CAS Number:  8012-89-3
Price
484,65 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Wax, bees
  • CAS Number:8012-89-3
  • MDL Number:MFCD00132754
  • Melting Pt:∼64 °C
  • Density:0.95 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-383-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.