Benzoic acid
25 kg
Catalog # ROTH5781.5
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:65-85-0
- Formula:C₇H₆O₂
- MDL Number:MFCD00002398
- Molecular Weight:122.12 g/mol
- Merck Index:13,01092
- Boiling Pt:249 °C (1013 hPa)
- Melting Pt:121…123 °C
- Density:1.316 g/cm³ (20 °C)
- Flash Pt:121 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-618-2
- REACH:01-2119455536-33
Specifications
Specifications
- Pk:25 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



