Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Potassium acetate

5 kg
Catalog # ROTH4986.2
Marketsource
Supplier:   Roth Carl
Potassium acetate
Catalog # ROTH4986.2
Supplier:  Roth Carl
Marketsource
CAS Number:  127-08-2
Potassium acetate
Potassium acetate

5 kg

CAS Number:  127-08-2
Price
Inquire For Price
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Acetic acid potassium salt
  • CAS Number:127-08-2
  • Formula:CH₃COOK
  • MDL Number:MFCD00012458
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,07687
  • Melting Pt:304 °C
  • Density:1.57 g/cm³ (25 °C)
  • Flash Pt:>250 °C
  • Storage Temperature:Ambient
  • EINECS:204-822-2
Specifications

Specifications

  • Pk:
    5 kg

Specifications