Potassium acetate
Glass bottle for solids, 1 kg
Catalog # ROTH4986.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Acetic acid potassium salt
- CAS Number:127-08-2
- Formula:CH₃COOK
- MDL Number:MFCD00012458
- Molecular Weight:98.14 g/mol
- Merck Index:13,07687
- Melting Pt:304 °C
- Density:1.57 g/cm³ (25 °C)
- Flash Pt:>250 °C
- Storage Temperature:Ambient
- EINECS:204-822-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



