Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Fructose

D-(-)-Fructose

25 kg
Catalog # ROTH4981.3
Marketsource
Supplier:   Roth Carl
D-(-)-Fructose
Catalog # ROTH4981.3
Supplier:  Roth Carl
Marketsource
CAS Number:  57-48-7
D-(-)-Fructose
D-(-)-Fructose

25 kg

CAS Number:  57-48-7
Price
Inquire For Price
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specifications

  • Pk:
    25 kg

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.