n-Hexane
Glass bottle, 1 L
Catalog # ROTH4723.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–94.3 °C
- Density:0.659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Storage Temperature:Ambient
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



