Order Entry
Denmark
Orders LinkContactUsLinkComponent
n-Hexane

n-Hexane

10 L
Catalog # ROTH3907.3
Flammable
Health
Irritant
Environment
Marketsource
Supplier:   Roth Carl
n-Hexane
Catalog # ROTH3907.3
Supplier:  Roth Carl
Marketsource
CAS Number:  110-54-3
n-Hexane
n-Hexane

10 L

CAS Number:  110-54-3
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94.3 °C
  • Density:0.659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
Specifications

Specifications

  • Pk:
    10 L

Specifications