Order Entry
Denmark
Orders LinkContactUsLinkComponent
GC columns, Rt®-608 (packed)
1,5% Rt-608 with PE AutoSys column configuration, Capillary, 2,00 mm, 2 m
Catalog # REST80221-840
GC columns, Rt®-608 (packed)
Catalog # REST80221-840
Supplier:  
GC columns, Rt®-608 (packed)
GC columns, Rt®-608 (packed)
1,5% Rt-608 with PE AutoSys column configuration, Capillary, 2,00 mm, 2 m
Discontinued
Product Details & Documents
Optimised for pesticide analysis by US EPA Method 608.
Specifications
  • Column Format:
    Capillary
  • Ø int.:
    2,00 mm
  • Description:
    1,5% Rt-608 with PE AutoSys column configuration
  • Length:
    2 m
  • Seq:
    05