Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cycloheximide

Cycloheximide

Glass bottle for solids, 1 g
Catalog # ROTH8682.1
Toxic
Health
Environment
Marketsource
Supplier:   Roth Carl
Cycloheximide
Catalog # ROTH8682.1
Supplier:  Roth Carl
Marketsource
CAS Number:  66-81-9
Cycloheximide
Cycloheximide

Glass bottle for solids, 1 g

CAS Number:  66-81-9
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.