Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Wiring

Wiring

Ledning, 0,2 mm, Konstantan (Ni/Cu-legering)
Catalog # 761-0143
Marketsource
Wiring
Catalog # 761-0143
Marketsource
Wiring
Wiring

Ledning, 0,2 mm, Konstantan (Ni/Cu-legering)

Price
Inquire For Price
Log in for availability
1 * 100 m
Product Details & Documents

Bare wire in a roll, on a plastic former.

Specifications
  • Beskrivelse:
    Ledning, 0,2 mm
  • Materiale:
    Konstantan (Ni/Cu-legering)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.