Engine models
Stirling engine
Catalog # 765-0172
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
Product Details & Documents
Intended for use on overhead projectors for demonstration of function and principle for different types of motors. Made of strong, transparent plastic. Equipped with suction feet.
- Dimensions: 250 × 250 mm
Specifications
- Description:Stirling engine