Modular diode laser
Modular diode laser, 635 nm, 3 mW
Catalog # 765-1004
Jump to:
Discontinued
Product Details & Documents
The advantages of a diode laser compared to a HeNe laser are mainly small format, low power consumption and long service life, as well as the possibility of electronic modulation. The laser is modular in the range 100 Hz 20 MHz. Dimensions.
- Elastic in width and thus easy to apply
Specifications
- Description:Modular diode laser, 635 nm, 3 mW