Order Entry
Denmark
Orders LinkContactUsLinkComponent
Guanine

Guanine

Glass bottle for solids, 10 g
Catalog # RESERES5137G-A101X
Supplier:   RESEARCH ORGANICS
Guanine
Catalog # RESERES5137G-A101X
Supplier:  RESEARCH ORGANICS
CAS Number:  73-40-5
Guanine

Glass bottle for solids, 10 g

CAS Number:  73-40-5
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  2-Amino-1H-purin-6(7H)-one
  • CAS Number:73-40-5
  • Formula:C₅H₅N₅O
  • MDL Number:MFCD00071533
  • Molecular Weight:151.13 g/mol
  • Merck Index:13,04580
  • Melting Pt:> 220 °C
  • Density:1.67 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-799-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.