Order Entry
Denmark
Orders LinkContactUsLinkComponent
MINICYL regulators

MINICYL regulators

MINICYL regulator, 3,2 mm brass fittings, max pressure: 300 psi
Catalog # REST20610
Supplier:   Restek
MINICYL regulators
Catalog # REST20610
Supplier:  Restek
MINICYL regulators
MINICYL regulators

MINICYL regulator, 3,2 mm brass fittings, max pressure: 300 psi

Price
2.070,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

This compact general purpose regulator is constructed of lightweight aluminum with an elastomer diaphragm. It has many laboratory applications including air-drying glassware, sparging or evaporating solutions and controlling pneumatic valves.

Specifikationer
  • Description:
    MINICYL regulator, 3,2 mm brass fittings, max pressure: 300 psi
  • Seq:
    01