Order Entry
Denmark
Orders LinkContactUsLinkComponent
Fingertight capillary column nuts
Fingertight capillary column nuts
Finger-tight capillary column nut, for use with standard 1,59 mm-type ferrules
Catalog # REST21312
Supplier:   Restek
Fingertight capillary column nuts
Catalog # REST21312
Supplier:  Restek
Fingertight capillary column nuts
Fingertight capillary column nuts
Finger-tight capillary column nut, for use with standard 1,59 mm-type ferrules
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These fingertight capillary column nuts rapidly tighten without wrenches and can be used with 0,25, 0,32, or 0,53 mm ID columns.
Specifikationer
  • Description:
    Finger-tight capillary column nut, for use with standard 1,59 mm-type ferrules
  • Seq:
    02