Storage Boxes
Trays, Red, 312×427×75 mm
Catalog # 763-0302
Supplier: GRATNELL'S
Jump to:
Product Details & Documents
Polypropylene storage trays. The material is fire-resistant and resistant to most chemicals in the school environment. Stackable.
- BSI tested and passed for heavy educational use
- All trays fit standard school furniture
- Lids and inserts also available
- Fully recyclable
Specifications
- Colour:Red
- Description:Trays
- W×D×H:312×427×75 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



