Order Entry
Denmark
Orders LinkContactUsLinkComponent
Storage trolleys

Storage trolleys

Double trolley without trays, 710×420×850 mm
Catalog # 765-0936
Supplier:   GRATNELL'S
Storage trolleys
Catalog # 765-0936
Supplier:  GRATNELL'S
Storage trolleys
Storage trolleys

Double trolley without trays, 710×420×850 mm

Price
4.190,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Flexible and easy-to-work carts. Stand of 25 mm square steel tube. Coating of white resistant epoxy paint. Heavy-duty nylon-coated wheels for quiet and smooth running.

The wagons are loaded with varying numbers of trays depending on the subject. Trolley is available in three different lengths and sections.

Slopes and hill holders will be added.

Specifications
  • Description:
    Double trolley without trays
  • Seq:
    02
  • W×D×H:
    710×420×850 mm