Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Storage trolleys

Single trolley with six trays, 370×420×850 mm
Catalog # 765-0935
Supplier:   GRATNELL'S
Storage trolleys
Catalog # 765-0935
Supplier:  GRATNELL'S
Storage trolleys
Storage trolleys

Single trolley with six trays, 370×420×850 mm

Price
3.230,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Flexible and easy-to-work carts. Stand of 25 mm square steel tube. Coating of white resistant epoxy paint. Heavy-duty nylon-coated wheels for quiet and smooth running.

The wagons are loaded with varying numbers of trays depending on the subject. Trolley is available in three different lengths and sections.

Slopes and hill holders will be added.

Specifikationer
  • Description:
    Single trolley with six trays
  • Seq:
    01
  • W×D×H:
    370×420×850 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.