Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Labeling tapes, TX

Label printer tape, Polyester, Thermal transfer, Black/red, PT-7000/8000, 12 mm×15 m
Catalog # BROTTX-431
Supplier:   BROTHER
Labeling tapes, TX
Catalog # BROTTX-431
Supplier:  BROTHER
Labeling tapes, TX
Labeling tapes, TX

Label printer tape, Polyester, Thermal transfer, Black/red, PT-7000/8000, 12 mm×15 m

Price
936,00 kr
Log in for availability
1 * 1 ROL
Product Details & Documents
Specifications
  • Printer model:
    PT-7000/8000
  • Printing method:
    Thermal transfer
  • Colour:
    Black/red
  • Description:
    Label printer tape
  • Dimensions:
    12 mm×15 m
  • Material:
    Polyester
  • Seq:
    07