Order Entry
Denmark
Orders LinkContactUsLinkComponent
Labeling tapes, TX

Labeling tapes, TX

Label printer tape, Polyester, Thermal transfer, Black/white, PT-7000/8000, 12 mm×15 m
Catalog # BROTTX-335
Supplier:   BROTHER
Labeling tapes, TX
Catalog # BROTTX-335
Supplier:  BROTHER
Labeling tapes, TX
Labeling tapes, TX

Label printer tape, Polyester, Thermal transfer, Black/white, PT-7000/8000, 12 mm×15 m

Price
1.040,00 kr
Log in for availability
1 * 1 ROL
Product Details & Documents
Specifications
  • Printer model:
    PT-7000/8000
  • Printing method:
    Thermal transfer
  • Colour:
    Black/white
  • Description:
    Label printer tape
  • Dimensions:
    12 mm×15 m
  • Material:
    Polyester
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.