Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stirring blades

Stirring blades

90 mm, 2,5 mm, 24 mm
Catalog # WITG1.496.090
Supplier:   witeg Labortechnik
Stirring blades
Catalog # WITG1.496.090
Supplier:  witeg Labortechnik
Stirring blades
Stirring blades

90 mm, 2,5 mm, 24 mm

Price
127,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stirring blades made of PTFE.

Specifications
  • Depth:
    2,5 mm
  • Width:
    90 mm
  • Height:
    24 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.