Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Automatic burettes, Dr Schilling pattern, Schellbach stripe

5 ml, 0,05 ml, 0,050 ± ml
  HECH40820005
Automatic burettes, Dr Schilling pattern, Schellbach stripe
  HECH40820005
Automatic burettes, Dr Schilling pattern, Schellbach stripe
Automatic burettes, Dr Schilling pattern, Schellbach stripe

5 ml, 0,05 ml, 0,050 ± ml

 
661,00 kr
   
1 * 1 Items
 

AR-GLAS®, class B.

  • Burettes are calibrated to deliver (marked 'Ex')
  • Schellbach stripe and fused graduation, faceted outlet tip
  • Improved special pinch cock with titration screw for precise regulation

Precisely calibrated, tolerance 1,5 times the official tolerance of class AS

 
  • Capacity:
    5 ml
  • Seq:
    01
  • Division:
    0,05 ml
  • Tolerance:
    0,050 ± ml
Accessories

 

  •  
    Description
    Capacity
    Division
     
     
  • HECH40821005
    Spare burette, AR®/soda glass
    5 ml
    0,05 ml
       
    1 * 1 Items - 400,00 kr

 

  •  
    Description
    Capacity
    Division
     
     
  • HECH40823001
    Spare plastic base, size 1, for 500 ml Schilling bottles
    -
    -
       
    1 * 1 Items - 47,80 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.