Pipette tips
Protectainer™, 1250 µl, -, Bulk, 750 tips
Catalog # VIST4026
Supplier: VISTALAB TECHNOLOGIES
Jump to:
Product Details & Documents
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Specifications
- Packed:Bulk, 750 tips
- Sterile:-
- Volume:1250 µl
- Seq:11
- Tip style:Protectainer™