Order Entry
Denmark
Orders LinkContactUsLinkComponent
Erlenmeyer flasks with screw caps

Erlenmeyer flasks with screw caps

100 ml, 105 mm, 64 mm, GL 25
Catalog # WITG5.507.003
Supplier:   witeg Labortechnik
Erlenmeyer flasks with screw caps
Catalog # WITG5.507.003
Supplier:  witeg Labortechnik
Erlenmeyer flasks with screw caps
Erlenmeyer flasks with screw caps

100 ml, 105 mm, 64 mm, GL 25

Price
221,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Erlenmeyer flasks, made of borosilicate glass, with screw-thread, graduated can be used for the storage of culture media and mixing.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Specifications
  • Ext.Ø:
    64 mm
  • Thread:
    GL 25
  • Capacity:
    100 ml
  • Height:
    105 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.