Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Acrylamide

Glass bottle for solids, 1,5 kg
Catalog # SHETIB70026
Toxic
Health
Supplier:   IBI SCIENTIFIC
Acrylamide
Catalog # SHETIB70026
Supplier:  IBI SCIENTIFIC
CAS Number:  79-06-1
Acrylamide
Acrylamide

Glass bottle for solids, 1,5 kg

CAS Number:  79-06-1
Price
13.110,00 kr
Log in for availability
1 * 1,5 kg
Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:232 °C (1013 hPa)
  • Melting Pt:84.5 °C
  • Density:1.122 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1,5 kg

Specifications