Comparator kits for water testing
Molybdate Test Kit, Model MO-2, 0 - 0,8/0 - 40 mg/l, 100, 1 KIT
Catalog # HACH1419301
Supplier: Hach
Jump to:
Product Details & Documents
Qualitative results using traditional colour comparator disc methods.
- Portable - ideal for field testing
- Simple fast method
- Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
- Pk:1 KIT
- Description:Molybdate Test Kit, Model MO-2
- Tests per kit:100
- Range:0 - 0,8/0 - 40 mg/l
Specifications
Accessories
Product Information
- Catalog #PkAvailabilityPrice
- Ammonium molybdate reagent for phosphate determinations, 100 mlAmmonium molybdate reagent for phosphate determinations, 100 mlHACH11032H1Log in for availability1 * 1 Items - 240,00 kr