Glycine
Glass bottle for solids, 500 g
RESERES5037G-A102X
: Aminoacetic acid, Glycocoll, H-Gly-OH
- :56-40-6
- :NH₂CH₂COOH
- :MFCD00008131
- :75.07 g/mol
- :13,04504
- :181 °C (1013 hPa)
- :83 °C
- :1.64 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ300
- :Ambient
- :200-272-2
- Pack type:Glass bottle for solids
- Pk:500 g