Order Entry
Denmark
Orders LinkContactUsLinkComponent
Glycine

Glycine

Glass bottle for solids, 100 g
Catalog # RESERES5037G-A101X
Supplier:   RESEARCH ORGANICS
Glycine
Catalog # RESERES5037G-A101X
Supplier:  RESEARCH ORGANICS
CAS Number:  56-40-6
Glycine
Glycine

Glass bottle for solids, 100 g

CAS Number:  56-40-6
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS Number:56-40-6
  • Formula:NH₂CH₂COOH
  • MDL Number:MFCD00008131
  • Molecular Weight:75.07 g/mol
  • Merck Index:13,04504
  • Boiling Pt:181 °C (1013 hPa)
  • Melting Pt:83 °C
  • Density:1.64 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:200-272-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.