Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

2-Mercaptoethanol

Glass bottle, 250 mL
Catalog # ROTH4227.1
Toxic
Corrosive
Environment
Marketsource
Supplier:   Roth Carl
2-Mercaptoethanol
Catalog # ROTH4227.1
Supplier:  Roth Carl
Marketsource
CAS Number:  60-24-2
2-Mercaptoethanol
2-Mercaptoethanol

Glass bottle, 250 mL

CAS Number:  60-24-2
Price
Inquire For Price
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  2-Hydroxyethyl mercaptan, β-Mercaptoethanol
  • CAS Number:60-24-2
  • Formula:HSCH₂CH₂OH
  • MDL Number:MFCD00004890
  • Molecular Weight:78.14 g/mol
  • Merck Index:13,05894
  • Boiling Pt:157 °C (1013 hPa)
  • Melting Pt:–100 °C
  • Density:1.12 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:200-464-6
  • UN:2966
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications