1-Tetradecanol
Glass bottle, 250 mL
Catalog # ROTH4741.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol
- CAS Number:112-72-1
- Formula:H₃C(CH₂)₁₂CH₂OH
- MDL Number:MFCD00004757
- Molecular Weight:214.39 g/mol
- Merck Index:13,06361
- Boiling Pt:289 °C (1013 hPa)
- Melting Pt:37…39 °C
- Density:0,8 g/cm³ (60 °C)
- Storage Temperature:Stue temperatur
- EINECS:204-000-3
Specifications
Specifications
- Pakning:Glass bottle
- Pakn:250 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



