Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
White modules, framed, C8, PolySorp surface, White, 96
738-0752
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
738-0752
PS, 96 wells per frame. For fluorescence/luminescence-based immunoassays and binding assays, these modules have been designed using the standard 96 MicroWell™ LockWell™ format. Opaque white modules have a reflective surface and offer low cross talk and minimal background. Opaque black modules with light absorbing surface offer minimum cross talk and back scattered light.
- Each well locks in the frame of the same height to prevent jamming in machines
- Easy to handle
- Uniform washing and reading
- Certified binding homogeneity for all four surfaces
- MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains; PolySorp™ surface has a high affinity for hydrophobic molecules
Total volume per well: 350 µl
- No. of wells:96
- Colour:White
- Description:White modules, framed, C8, PolySorp surface
- Cat. No.:738-0752
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact