Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Closures for test tubes and flasks

Cotton cheesecloth, grade 50, 24 - 28 mm
  734-1264
Closures for test tubes and flasks
  734-1264
Closures for test tubes and flasks
Closures for test tubes and flasks

Cotton cheesecloth, grade 50, 24 - 28 mm

 
12.970,00 kr
   
1 * 700 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
 

Identy-plug.

Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.

Cotton cheesecloth.

100% pure cotton, very absorbent and low linting; 28×24 thread count.

Bleached cotton balls.

Available large or medium.

 
  • Description:
    Cotton cheesecloth, grade 50, 24 - 28 mm
  • Seq:
    07