Order Entry
Denmark
Orders LinkContactUsLinkComponent
Super syringes
Super syringes
Super syringe with TLL connector, 2 l, S2000 TLL
Catalog # HAMI86314
Supplier:   HAMILTON BONADUZ
Super syringes
Catalog # HAMI86314
Supplier:  HAMILTON BONADUZ
Super syringes
Super syringes
Super syringe with TLL connector, 2 l, S2000 TLL
Price
15.020,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Super syringes with PTFE Luer lock (TLL) connector or tracheal adapter.
  • Clear acrylic syringe barrels
  • For use with gases and liquids
Ideal for use with air sampling, preparing gas standards, calibrating reservoirs, and pneumographs.
Specifications
  • Type:
    S2000 TLL
  • Volume:
    2 l
  • Description:
    Super syringe with TLL connector
  • Seq:
    04
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • TLL connector
    TLL connector
    HAMI18026
    super syringe S0500, S1000, S1500, S2000
    Log in for availability
    1 * 1 Items - 656,00 kr