Super syringes
Super syringe with TLL connector, 0,5 l, S0500 TLL
Catalog # HAMI86311
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
Super syringes with PTFE Luer lock (TLL) connector or tracheal adapter.
- Clear acrylic syringe barrels
- For use with gases and liquids
Ideal for use with air sampling, preparing gas standards, calibrating reservoirs, and pneumographs.
Specifications
- Type:S0500 TLL
- Volume:0,5 l
- Description:Super syringe with TLL connector
- Seq:01
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- TLL connectorTLL connectorHAMI18026super syringe S0500, S1000, S1500, S2000Log in for availability1 * 1 Items - 656,00 kr