Order Entry
Denmark
Orders LinkContactUsLinkComponent
Fingertight adapters, Color-It
Fingertight adapters, Color-It
Assorted
Katalog # JOURJR-55016-12
Leverandør:   VICI JOUR RESEARCH
Fingertight adapters, Color-It
Katalog # JOURJR-55016-12
Leverandør:  VICI JOUR RESEARCH
Fingertight adapters, Color-It
Assorted
Price
217,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents
These fingertight PP adapters are used with ¹/₄" PEEK and stainless hex-head nuts, converting them into colour coded fingertight fittings.
  • Fit all hex-head PEEK and SS fittings for ¹/₁₆" tubing
  • Used in low and high pressure
  • Available in six colours for easy solvent line identification
Specifikationer
  • Colour:
    Assorted
  • Seq:
    07